KEY466132635 Key Organics / BIONET

4-[(4-methoxyphenyl)methyl]-8-(4-nitrophenyl)-1-oxa-4,8-diazaspiro[4.5]decane

  • MDL number MFCD00173321
  • Cas Number : 303151-64-6
  • Molecular Weight : 383.448

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4620

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11G-938_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart