KEY466132641 Key Organics / BIONET

3-{3-[(3-nitropyridin-2-yl)sulfanyl]-1H-1,2,4-triazol-1-yl}butan-2-one

  • MDL number MFCD00794528
  • Cas Number : 303151-95-3
  • Molecular Weight : 293.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4662

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart