KEY466132646 Key Organics / BIONET

4-[2-benzoyl-5-(trifluoromethyl)pyridin-3-yl]morpholine

  • MDL number MFCD00244623
  • Cas Number : 303151-96-4
  • Molecular Weight : 336.314

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4671

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-042_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart