KEY466132672 Key Organics / BIONET

N-(2,2-diethoxyethyl)-3-(2-fluoro-4-nitrophenoxy)thiophene-2-carboxamide

  • MDL number MFCD00215001
  • Cas Number : 303152-59-2
  • Molecular Weight : 398.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4747

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-386S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart