KEY466132683 Key Organics / BIONET

2-[(3-chlorophenyl)amino]-4H-3,1-benzoxazin-4-one

  • MDL number MFCD00262915
  • Cas Number : 81905-03-5
  • Molecular Weight : 272.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4776

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-537S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart