KEY466132688 Key Organics / BIONET

ethyl 1'-(pyrimidin-2-yl)-3a,4,8,8a-tetrahydrospiro[[1,3]dioxepino[5,6-d][1,2]oxazole-6,4'-piperidine]-3-carboxylate

  • MDL number MFCD00664177
  • Cas Number : 303152-98-9
  • Molecular Weight : 362.386

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4800

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-921_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart