KEY466132689 Key Organics / BIONET

1'-(pyrimidin-2-yl)-3a,4,8,8a-tetrahydrospiro[[1,3]dioxepino[5,6-d][1,2]oxazole-6,4'-piperidine]-3-carbohydrazide

  • MDL number MFCD00664179
  • Cas Number : 303153-00-6
  • Molecular Weight : 348.363

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-924_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart