KEY466132691 Key Organics / BIONET

(2Z)-3-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazin-1-yl}-2-cyanobut-2-enamide

  • MDL number MFCD00663784
  • Cas Number : 303153-04-0
  • Molecular Weight : 333.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4807

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-932_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart