KEY466132709 Key Organics / BIONET

2-(3,4-dichlorobenzoyl)-3-methyl-5-nitro-1-benzofuran

  • MDL number MFCD00665526
  • Cas Number : 400076-04-2
  • Molecular Weight : 350.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4971

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11J-545S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart