KEY466132712 Key Organics / BIONET

2-benzoyl-6-methoxy-3-methyl-1-benzofuran

  • MDL number MFCD00243858
  • Cas Number : 269075-46-9
  • Molecular Weight : 266.296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4974

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11J-549S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart