KEY466132725 Key Organics / BIONET

methyl 1-[5-(3-chlorothiophen-2-yl)-3-cyano-1H-pyrrol-2-yl]piperidine-4-carboxylate

  • MDL number MFCD00794209
  • Cas Number : 477854-08-3
  • Molecular Weight : 349.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5009

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11J-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart