KEY466132730 Key Organics / BIONET

5-(4-fluorophenyl)-4H,5H-thieno[2,3-d]pyridazine

  • MDL number MFCD00232237
  • Cas Number : 303145-59-7
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357967

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart