KEY466132733 Key Organics / BIONET

3-(4-methylphenyl)-5H-indeno[1,2-c]pyridazin-5-one

  • MDL number MFCD00232445
  • Cas Number : 300843-87-2
  • Molecular Weight : 272.307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357973

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart