KEY466132740 Key Organics / BIONET

6-(4-chlorophenyl)-[1,2,4]triazolo[1,5-a]pyrimidine-2,7-diamine

  • MDL number MFCD00232455
  • Cas Number : 303145-69-9
  • Molecular Weight : 260.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357985

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart