KEY466132748 Key Organics / BIONET

methyl 6-chloro-5-cyano-2-methylpyridine-3-carboxylate

  • MDL number MFCD01315275
  • Cas Number : 303146-25-0
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5111

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-301S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart