KEY466132758 Key Organics / BIONET

(5E)-6-(4-ethoxyphenyl)-4-oxohex-5-enoic acid

  • MDL number MFCD04124280
  • Cas Number : 121365-28-4
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5142

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-347S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart