KEY466132760 Key Organics / BIONET

(5E)-4-oxo-6-[4-(propan-2-yl)phenyl]hex-5-enoic acid

  • MDL number MFCD01315276
  • Cas Number : 303146-58-9
  • Molecular Weight : 246.306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5152

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-363S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart