KEY466132766 Key Organics / BIONET

6-[(E)-2-phenylethenyl]-2-{[3-(trifluoromethyl)phenyl]methyl}-2,3,4,5-tetrahydropyridazin-3-one

  • MDL number MFCD00793321
  • Cas Number : 303146-66-9
  • Molecular Weight : 358.364

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5160

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-371S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart