KEY466132769 Key Organics / BIONET

2-[(3,4-dichlorophenyl)methyl]-6-[(E)-2-phenylethenyl]-2,3,4,5-tetrahydropyridazin-3-one

  • MDL number MFCD00793325
  • Cas Number : 1164529-92-3
  • Molecular Weight : 359.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-381S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart