KEY466132793 Key Organics / BIONET

1-[2-(3,4-dimethoxyphenyl)ethyl]-6-methyl-4-oxo-1,4-dihydropyrimidine-5-carbonitrile

  • MDL number MFCD00663798
  • Cas Number : 303148-43-8
  • Molecular Weight : 299.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5314

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-919_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart