KEY466132797 Key Organics / BIONET

2-methyl-3-[2-oxo-2-(thiophen-2-yl)ethyl]-1,4-dihydronaphthalene-1,4-dione

  • MDL number MFCD00243895
  • Cas Number : 130836-94-1
  • Molecular Weight : 296.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5323

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-929_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart