KEY466132800 Key Organics / BIONET

N'-[(1E)-1-(4-fluorophenyl)ethylidene]methoxycarbohydrazide

  • MDL number MFCD00792964
  • Cas Number : 112382-27-1
  • Molecular Weight : 210.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5330

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart