KEY466132816 Key Organics / BIONET

(3Z)-3-[2-(4-fluorophenyl)hydrazin-1-ylidene]-1-phenyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD00975809
  • Cas Number : 303984-91-0
  • Molecular Weight : 331.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5421

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-391S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart