KEY466132825 Key Organics / BIONET

2-{[(2-chlorophenyl)methyl]sulfanyl}-4-hydroxy-6-phenylpyrimidine-5-carbonitrile

  • MDL number MFCD00975818
  • Cas Number : 303985-05-9
  • Molecular Weight : 353.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5441

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-521S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart