KEY466132830 Key Organics / BIONET

(3Z)-1-[(3,4-dichlorophenyl)methyl]-3-(2-phenylhydrazin-1-ylidene)-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD00975829
  • Cas Number : 303985-17-3
  • Molecular Weight : 396.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5468

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-564S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart