KEY466132838 Key Organics / BIONET

2-{[(4-methoxyphenyl)methyl]sulfanyl}-N-methyl-6-[(methylsulfanyl)methyl]pyrimidin-4-amine

  • MDL number MFCD00975734
  • Cas Number : 303985-38-8
  • Molecular Weight : 321.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-601S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart