KEY466132851 Key Organics / BIONET

N-(2-benzoyl-4-chlorophenyl)-3-chloro-2,2-dimethylpropanamide

  • MDL number MFCD00793378
  • Cas Number : 303985-83-3
  • Molecular Weight : 350.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5547

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart