KEY466132853 Key Organics / BIONET

(4E)-4-[2-(3-nitrophenyl)hydrazin-1-ylidene]-3,4-dihydro-1H-2-benzopyran-1,3-dione

  • MDL number MFCD00793380
  • Cas Number : 303985-86-6
  • Molecular Weight : 311.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5551

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart