KEY466132866 Key Organics / BIONET

2-(4-chlorophenyl)-6-[(2E)-3-(dimethylamino)prop-2-enoyl]-4-methyl-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD00794221
  • Cas Number : 1164529-87-6
  • Molecular Weight : 334.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5612

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-926_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart