KEY466132869 Key Organics / BIONET

2-(1,3-dioxo-2,3-dihydro-1H-inden-2-ylidene)-2-{[(pyridin-2-yl)methyl]amino}acetonitrile

  • MDL number MFCD00974975
  • Cas Number : 117457-21-3
  • Molecular Weight : 289.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5641

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-958_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart