KEY466132884 Key Organics / BIONET

2-{2-[(1E)-[(benzyloxy)imino]methyl]-1H-pyrrol-1-yl}-4-(dimethylamino)pyridine-3-carbonitrile

  • MDL number MFCD01314676
  • Cas Number : 303986-48-3
  • Molecular Weight : 345.406

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5672

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart