KEY466132903 Key Organics / BIONET

5-[(4-methoxyphenyl)methyl]-4-oxo-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2-carboxylic acid

  • MDL number MFCD02083066
  • Cas Number : 477854-40-3
  • Molecular Weight : 301.302

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5714

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-319S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart