KEY466132916 Key Organics / BIONET

2-phenyl-4-{[3-(trifluoromethyl)phenyl]sulfanyl}pyrimidine-5-carboxylic acid

  • MDL number MFCD02083177
  • Cas Number : 477854-57-2
  • Molecular Weight : 376.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5759

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-522S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart