KEY466132921 Key Organics / BIONET

4-[(4-methoxyphenyl)sulfanyl]-2-phenylpyrimidine-5-carboxylic acid

  • MDL number MFCD02083183
  • Cas Number : 477854-63-0
  • Molecular Weight : 338.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5766

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-535S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart