KEY466132923 Key Organics / BIONET

4-[(2,5-dimethylphenyl)sulfanyl]-2-phenylpyrimidine-5-carboxylic acid

  • MDL number MFCD27924200
  • Cas Number : 1797376-50-1
  • Molecular Weight : 336.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264 - 272 (dec)
Purity 97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402870

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-537S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart