KEY466132940 Key Organics / BIONET

(4E)-2-[(2-chloro-6-fluorophenyl)methyl]-5-(dimethylamino)-1-phenylpent-4-ene-1,3-dione

  • MDL number MFCD06496102
  • Cas Number : 900015-47-6
  • Molecular Weight : 359.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-708_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart