KEY466132942 Key Organics / BIONET

(3Z)-1,1,1-trifluoro-4-[(4-methoxyphenyl)amino]pent-3-en-2-one

  • MDL number MFCD01568218
  • Cas Number : 201659-55-4
  • Molecular Weight : 259.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5806

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart