KEY466132952 Key Organics / BIONET

2-(naphthalene-2-sulfonyl)-3-phenyl-2,5-dihydro-1,2-oxazol-5-one

  • MDL number MFCD01568228
  • Cas Number : 303987-08-8
  • Molecular Weight : 351.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5836

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-921_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart