KEY466132967 Key Organics / BIONET

N-(2,5-difluorophenyl)-4-hydroxy-2-oxo-1-azatricyclo[7.3.1.0^{5,13}]trideca-3,5(13),6,8-tetraene-3-carboxamide

  • MDL number MFCD01568730
  • Cas Number : 303987-25-9
  • Molecular Weight : 356.329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5868

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11N-034_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart