KEY466132989 Key Organics / BIONET

2-(methylsulfanyl)-5,6,7,8-tetrahydroquinazolin-4-yl 2-chlorobenzoate

  • MDL number MFCD01568950
  • Cas Number : 303987-61-3
  • Molecular Weight : 334.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5972

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11N-355S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart