KEY466133008 Key Organics / BIONET

N-(4-fluorophenyl)-4-hydroxy-1-methyl-2,2-dioxo-1H-2lambda6-thieno[3,2-c][1,2]thiazine-3-carboxamide

  • MDL number MFCD01569071
  • Cas Number : 303987-81-7
  • Molecular Weight : 354.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11N-404S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart