KEY466133028 Key Organics / BIONET

4-[(4-methylphenyl)sulfanyl]-3-nitro-N-(propan-2-yl)benzamide

  • MDL number MFCD01569098
  • Cas Number : 303988-16-1
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6085

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11N-661S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart