KEY466133046 Key Organics / BIONET

ethyl 2-{2-amino-[1,2,4]triazolo[1,5-a]pyrimidin-7-yl}-3-(2-chloro-6-fluorophenyl)propanoate

  • MDL number MFCD01871427
  • Cas Number : 477856-20-5
  • Molecular Weight : 363.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6148

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11N-768_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart