KEY466133055 Key Organics / BIONET

2-[(4-chlorophenyl)amino]-4-(3-cyanophenyl)-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

  • MDL number MFCD27924201
  • Cas Number : 1347820-42-1
  • Molecular Weight : 347.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402871

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11N-909_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart