KEY466133070 Key Organics / BIONET

ethyl (2E)-2-chloro-2-[2-(2-nitrophenyl)hydrazin-1-ylidene]acetate

  • MDL number MFCD00505295
  • Cas Number : 37522-26-2
  • Molecular Weight : 271.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6235

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart