KEY466133073 Key Organics / BIONET

methyl 2-{4'-pentyl-[1,1'-biphenyl]-4-yl}acetate

  • MDL number MFCD02081963
  • Cas Number : 477856-65-8
  • Molecular Weight : 296.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6245

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart