KEY466133076 Key Organics / BIONET

1-(2,4-dichlorophenyl)-2-{[5-(4-methoxyphenyl)-1,3,4-oxadiazol-2-yl]sulfanyl}ethan-1-one

  • MDL number MFCD02186976
  • Cas Number : 477856-68-1
  • Molecular Weight : 395.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6254

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart