KEY466133083 Key Organics / BIONET

1-(4-chlorophenyl)-2-{[5-(pyridin-4-yl)-1,3,4-oxadiazol-2-yl]sulfanyl}ethan-1-one

  • MDL number MFCD01847740
  • Cas Number : 302552-42-7
  • Molecular Weight : 331.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6274

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-109_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart