KEY466133084 Key Organics / BIONET

methyl 2-{4-[4-(4-fluorophenyl)pyrimidin-2-yl]phenoxy}propanoate

  • MDL number MFCD02187277
  • Cas Number : 477856-80-7
  • Molecular Weight : 352.365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6277

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-114_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart