KEY466133085 Key Organics / BIONET

methyl 4-{4-[4-(4-fluorophenyl)pyrimidin-2-yl]phenoxy}butanoate

  • MDL number MFCD02187278
  • Cas Number : 477856-81-8
  • Molecular Weight : 366.392

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6279

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-116_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart